Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK11 Monoclonal Antibody (OTI4H5), TrueMAB™, OriGene
SDP

Catalog No. p-200009836
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2173 30 μL
50-167-2720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2173 Supplier OriGene Supplier No. TA501583S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK11 Monoclonal antibody specifically detects NEK11 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen NEK11
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4H5
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FSHB
Gene Accession No. P01225
Gene Alias 4932416N14Rik; Nek11; never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA (never in mitosis gene a)-related expressed kinase 11; NIMA related kinase 11; NIMA-related kinase 11; nimA-related protein kinase 11; Serine/threonine-protein kinase Nek11
Host Species Mouse
Immunogen Full length human recombinant protein of human FSHB produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.