Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104126 Supplier Thermo Scientific Supplier No. PA5104126
Only null left
Add to Cart
Add to Cart

NEK4 Polyclonal Antibody

NEK4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NEK4
Gene Accession No. P51957, Q9Z1J2
Gene Alias Nek4; Never in mitosis A-related kinase 4; NIMA (never in mitosis gene a)-related expressed kinase 4; NIMA (never in mitosis gene a)-related kinase 4; NIMA related kinase 4; NIMA-related kinase 4; nimA-related protein kinase 4; NRK2; pp12301; serine/threonine kinase 2; serine/threonine protein kinase-2; serine/threonine-protein kinase 2; Serine/threonine-protein kinase Nek4; serine/threonine-protein kinase Nek4 variant 2; serine/threonine-protein kinase NRK2; STK2
Gene Symbols NEK4
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEK4(Accession P51957), corresponding to amino acid residues H499-E549.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23955, 306252, 6787
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.