Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nelfinavir Mesylate Hydrate 97.0+%, TCI America™
SDP

Catalog No. N0986100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0986100MG 100 mg
N098625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0986100MG Supplier TCI America Supplier No. N0986100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 159989-65-8
Molecular Formula C33H49N3O7S2
Molecular Weight (g/mol) 663.889
MDL Number MFCD00931436
InChI Key NQHXCOAXSHGTIA-SKXNDZRYSA-N
Synonym nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777
PubChem CID 64142
ChEBI CHEBI:7497
IUPAC Name (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid
SMILES CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 663.89
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nelfinavir Mesylate Hydrate
RTECSNumber NW6995333
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.