Learn More
United States Biological Corporation Nematode Growth Medium, Lite (NGM Lite), Powder, 500g
Catalog No. NC9379170 Shop All United States Biological Corporation Products
Encompass_Preferred
A modified preparation of NGM for use in Nematode growth studies. Appearance: Light tan, homogeneous, free flowing powder Solubility without autoclaving: Light tan, incomplete Solubility with autoclaving: Light tan, clear, complete pH (after autoclaving): 5.5-6.5
Catalog No. NC9379170 Supplier United States Biological Corporation Supplier No. N1005500G
A modified preparation of NGM for use in Nematode growth studies. Appearance: Light tan, homogeneous, free flowing powder Solubility without autoclaving: Light tan, incomplete Solubility with autoclaving: Light tan, clear, complete pH (after autoclaving): 5.5-6.5