Learn More
Ansell Nemesis™ Polarized Safety Eyewear
Catalog No. 19066118 Shop All Ansell Products
Change view
Click to view available options
Lens Color: Smoke; Lens Type: Polarized; Frame Color: Camo
Catalog No.
19-066-118 Lens Color: Smoke; Lens Type: Polarized; Frame Color: Camo
1 options
This item is not returnable. View return policy
Catalog No. 19-066-118 Supplier Ansell Supplier No. 47417
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Polarized-lens technology to reduce eye stress and fatigue caused by excessive glare
  • Sporty, flexible, lightweight
  • Reduce eye stress and fatigue caused by excessive glare
  • Designed to help reduce reflected glare, eye strain, eye fatigue
  • Anti-scratch lens coating increases lens longevity
  • EN 166 “K” mark testing for resistance to surface damage by fine particles
  • Polycarbonate material
  • 99.9% UVA/UVB/UVC protection
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Type Polarized
Certifications/Compliance ANSI Z87.1
Lens Tint Smoke
Product Type Protective Glasses
For Use With (Application) Viewing at indoors
Frame Color Camouflage
Includes Neck cord
Lens Material Polycarbonate
Frame Material Metallic
UV Protection UVA, UVB, UVC