Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEMF Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571157
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1157 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1157 Supplier Bethyl Laboratories, Inc Supplier No. A305757AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Nuclear export mediator factor, NEMF, plays a role in nuclear export. It is expressed in brain, heart, liver, lung, spleen, and skeletal muscle tissue, and expressed at lower levels in stomach and testis. It interacts (via its N-terminus) with XPO1; the N-terminal domain contains a nuclear export signal and the C-terminal domain may interact with cargo proteins. This protein is active in normal lung tissue but is inactive in several lung cancer cell lines.

Specifications

Antigen NEMF
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NEMF
Gene Accession No. O60524
Gene Alias Nuclear export mediator factor NEMF, Antigen NY-CO-1, Serologically defined colon cancer antigen 1, NEMF, SDCCAG1
Gene Symbols NEMF
Host Species Rabbit
Immunogen Between 950 and 1000
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9147
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.