Learn More
Electron Microscopy Sciences Neo Micro Coverglass 24 x 50 mm
Supplier: Electron Microscopy Sciences 7220720
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly.