Learn More
AdipoGen Neocuproine hemihydrate
SDP
Supplier:  AdipoGen CDXN0115G100
Encompass_Preferred
Chemical. CAS 484-11-7. Formula C14H12N2 . 0.5H2O. MW 217.27. Neocuproine is a complexing reagent, known for its ability to form stable complexes with metal ions, particularly CU2 ions. This property makes it valuable in various analytical and research applications, including spectrophotometry and electrochemistry. In the presence of CU2 ions, neocuproine forms a brightly colored complex that can be used for the quantitative determination of copper in solution. The Neocuproine-CU2 complex exists in a 21 ratio with a maximum absorption at 454 nm. Neocuproine is used in complex with Cu2 as chromogenic oxidant in the total antioxidant capacity assay CUPRAC method. In addition to its analytical uses, neocuproine and its derivatives have been employed in the field of coordination chemistry to study and synthesize metal complexes.
Catalog No. 50-256-0409
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.