Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neocuproine Hydrochloride Monohydrate 99.0+%, TCI America™
SDP

Catalog No. N042325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N042325G 25 g
N04231G 1 g
N04235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N042325G Supplier TCI America Supplier No. N042325G

Chemical Identifiers

CAS 303136-82-5
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00150062
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym 2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 %
PubChem CID 2723838
IUPAC Name 2,9-dimethyl-1,10-phenanthroline
SMILES CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 244.72
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Neocuproine Hydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.