Learn More
Specifications
Specifications
| Color | Colorless to Yellow |
| Boiling Point | 65°C |
| Melting Point | -98°C |
| CAS Max % | 99.9, 0.1 |
| Linear Formula | C14H14N2O |
| UN Number | UN1993 |
| Density | 0.79g/mL |
| Solubility Information | Soluble in water |
| Formula Weight | 226.27 |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Toxic if swallowed, in contact with skin or if inhaled. Causes damage to organs (central nervous system, kidneys, liver, optic nerve). |
| Precautionary Statement | GHS P Statement Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor. In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish. Store locked up in a cool, well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.