Learn More
Miele, Inc. Neodisher FLA 19L/25KG Liquid
Supplier:  Miele, Inc. 94000160
Encompass_Preferred
Neodisher FLA 19L/25KG LiquidFor the automated cleaning of laboratory glassware.Free of surfactants and active chlorine
Catalog No. 50-171-3115
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.