Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium standard solution, for AAS, 1mg/mL Nd in 2% HNO3, Thermo Scientific™

Catalog No. AC196145000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196145000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196145000 Supplier Thermo Scientific Chemicals Supplier No. 196145000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-00-8 , 7697-37-2 , 7732-18-5
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

CAS Min % 1.9, 96.9
CAS Max % 2.1, 97.1
Physical Form Solution
Concentration 995 to 1005 μg/mL of Nd in 2% HNO3 (20°C)
Quantity 500 mL
Formula Weight 144.24
Grade AAS
Chemical Name or Material Neodymium standard solution, 1 mg/ml Nd in 2% HNO3
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.