Learn More
Neogen Diagnostics K-Gold Plus PNPP Substrate, 1 L, Microwell, AP, Colorimetric
Supplier: Neogen Diagnostics 339177
K-GoldPlus is an innovative one-bottle p-nitrophenyl-phosphate (PNPP) substrate that offers excellent activity and signal-to-noise ratio compared to other PNPP substrates. K-Gold Plus offers 3040% rate enhancement compared to our original K-Gold substrate. This corresponds to an average increase in signal-to-background of 64% for K-Gold Plus. Stable for a minimum of 24-months when stored at 2-8C.