Learn More
Lkt Laboratories CAS 1405-10-3, ≥98%, Inhibitor of protein translation inhibitor, TRPV1 receptors, and P2X receptors.
SDP
Supplier:  Lkt Laboratories N1755
Encompass_Preferred
Neomycin is a polycationic aminoglycoside antibiotic that exhibits antibacterial and neuromodulatory activities; it was originally found in species of Streptomyces.  Neomycin binds to 23S rRNA, constraining tRNA in the P-site, inhibiting translocation and ribosomal recycling; together, this results in inhibition of protein synthesis. Neomycin B binds and inhibits bacterial RNase P and bacterial T box antiterminator RNA. Neomycin B inhibits mammalian RNA splicing as well. Neomycin also noncompetitively inhibits cation channels, including P2X receptors and transient receptor potential vanilloid receptor 1 (TRPV1) channels. Additionally, this compound inhibits Ca2+ influx, preventing presynaptic release of acetylcholine and norepinephrine in feline superior cervical ganglia.
Catalog No. NC2539825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.