Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Sulfate, MP Biomedicals™

Catalog No. MP218061005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218061005 5 g
MP218061001 1 g
MP218061010 10 g
MP218061025 25 g
MP218061080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP218061005 Supplier MP Biomedicals, Inc Supplier No. 0218061005
Add to Cart
Add to Cart

Acts against both gram-positive and gram-negative bacteria. Neomycin Sulfate, MP Biomedicals is produced by Streptomyces containing a minimum of 85% neomycin B.

This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Applications:

  • As a selection agent for prokaryotic cells transformed using the neo selectable marker gene
  • To study ototoxic side effects of antibiotics and platelet-derived growth factor responses in certain fibroblasts
  • In the extraction of Nuclear Phosphatidylinositol 4,5-Bisphosphate-Interacting Proteins
  • It is recommended for use in cell culture applications at 50mg/L
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
PubChem CID 124080932
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Physical Form Powder
pH 5 to 7.5
Loss on Drying <8.0%
Quantity 5 g
Solubility Information H2O soluble (50mg/mL)
IUPAC Name 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Formula Weight 908.9g/mol
Grade USP
Recommended Storage Room Temperature, desiccate, protect from light
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.