accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH2HOHONH2(S)(R)(S)O(R)OHH2NONH2OONH2HOHONH2OHOHOOHSOHOO

CAS RN 1405-10-3

IUPAC Name: 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Synonyms: 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Molecular Weight (g/mol): 712.72
Molecular Formula: C23H48N6O17S
InChi Key: OIXVKQDWLFHVGR-DJUBFIBWNA-N
SMILES: OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O
Physical Form 
  • (1)
  • (3)
  • (10)
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
  • (13)
Color 
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
Type 
  • (7)
  • (1)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (2)
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (4)
  • (5)
  • (6)
  • (1)
  • (3)
  • (10)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (13)
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (7)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)