Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Sulfate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S000015
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000015 100 g
S139821 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000015 Supplier G-Biosciences Supplier No. RC1064
Add to Cart
Add to Cart

Neomycin is a broad-spectrum aminoglycoside antibiotic isolated from Streptomyces species. It targets gram-negative and gram-positive bacteria.

Neomycin can also be used as a selection agent for prokaryotic cells transformed with the neo gene or NPT II resistance gene and to study toxic antibacterial side effects. In addition, neomycin is a calcium channel protein inhibitor and DNase I inhibitor. It can be used in a variety of cell culture applications

Chemical Identifiers

CAS 1405-10-3

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 100 g
Percent Purity 97%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.