Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Sulfate, USP, Spectrum™ Chemical
SDP

Catalog No. 18604770 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-770 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-770 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. NE11125KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

NE111, 1405-10-3, C23H46N6O13·3H2SO4·xH20

Spectrum™ Chemical Neomycin Sulfate, USP is an aminoglycoside antibiotic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
IUPAC Name 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Physical Form Powder
pH 5.0 to 7.5
CAS Max % 1
Loss on Drying 0.08
Packaging Fiber Drum
Color White or White/Yellow
Quantity 25 kg
Grade USP
Chemical Name or Material Neomycin Sulfate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.