Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neomycin Sulfate (White Powder), Fisher BioReagents

Catalog No. BP266925 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP266925 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP266925

missing translation for 'validMainframeMsg'

Neomycin Sulfate is an antibiotic and is used in tissue culture studies. It can act as an inhibitor of protein biosynthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
Synonym Neobiotic
PubChem CID 124080932
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Type Neomycin Sulfate
Physical Form Solid
pH 6.8
Loss on Drying < 8%
Packaging Amber Glass
Potency > 650μg/mg
Color White
Quantity 25 g
Assay Percent Range > 98.5 %
Merck Index 15, 6539
IUPAC Name (2S,3R,4S,5S,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6R)-4,6-diamino-2-[(2R,3S,4R,5S)-4-[(2S,3R,4S,5R,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid
Formula Weight 614.65
Percent Purity >98.5%
Chemical Name or Material Neomycin Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Substances known to cause developmental toxicity in humans. May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:1
Recommended Storage RT, desiccate, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.