Learn More
Enzo Life Sciences (+)-Neopterin (50mg). CAS: 2009-64-5
SDP
Supplier:  Enzo Life Sciences ALX550126M050
Encompass_Preferred
Biochemical marker used as an indicator of cell proliferation. Pterin cofactor. Precursor of tetrahydrobiopterin. Alternative name: D(+)-6-(erythro-1,2,3-Trihydroxypropyl)pterin. Purity: ≥98% (HPLC). Solubility: Soluble in 0.05M NaOH or 1M HCl; very slightly soluble in water. Ultrasonication may improve the dissolution. Long Term Storage: -20°C.
Catalog No. 50-200-9484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.