Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neostigmine Bromide 99.0+%, TCI America™
SDP

Catalog No. N035825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035825G 25 g
N03585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N035825G Supplier TCI America Supplier No. N035825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 114-80-7
Molecular Formula C12H19BrN2O2
Molecular Weight (g/mol) 303.20
MDL Number MFCD00011795
InChI Key LULNWZDBKTWDGK-UHFFFAOYSA-M
Synonym neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide
PubChem CID 8246
ChEBI CHEBI:179557
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 303.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Neostigmine Bromide
EINECSNumber (9)-0708&(9)-1092
RTECSNumber BR3150000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.