Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neostigmine Methyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. N044725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N044725G 25 g
N04475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N044725G Supplier TCI America Supplier No. N044725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-60-5
Molecular Formula C13H22N2O6S
Molecular Weight (g/mol) 334.387
MDL Number MFCD00011796
InChI Key OSZNNLWOYWAHSS-UHFFFAOYSA-M
Synonym (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulfate, (3-Hydroxyphenyl)trimethylammonium Methyl Sulfate Dimethylcarbamate
PubChem CID 5824
ChEBI CHEBI:7516
IUPAC Name [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;methyl sulfate
SMILES CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C.COS(=O)(=O)[O-]

Specifications

Melting Point 146°C
Color White
pH 3.0 - 5.0
Quantity 25 g
UN Number 2811
Formula Weight 334.39
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Neostigmine Methyl Sulfate
EINECSNumber (9)-0708
RTECSNumber CY1225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.