Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepafenac 98.0+%, TCI America™
SDP

Catalog No. N09321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09321G 1 g
N0932200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09321G Supplier TCI America Supplier No. N09321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78281-72-8
Molecular Formula C15H14N2O2
Molecular Weight (g/mol) 254.29
MDL Number MFCD08067732
InChI Key QEFAQIPZVLVERP-UHFFFAOYSA-N
Synonym 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide
PubChem CID 151075
ChEBI CHEBI:75922
IUPAC Name 2-(2-amino-3-benzoylphenyl)acetamide
SMILES NC(=O)CC1=CC=CC(C(=O)C2=CC=CC=C2)=C1N

Specifications

Melting Point 184°C
Color Yellow
Quantity 1 g
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nepafenac
RTECSNumber CY1480710
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.