Learn More
Apexbio Technology LLC Nepicastat (SYN-117) HCl,5mg CAS# 170151-24-3
SDP
Supplier:  Apexbio Technology LLC A43325
Encompass_Preferred
Nepicastat (SYN-117) HCl is a potent and selective inhibitor of dopamine-β-hydroxylase with IC50 values of 8.5 and 9.0 nM in bovine and human, respectively [1]. Dopamine-β-hydroxylase is an enzyme involved in the synthesis of small-molecule membrane-bound neurotransmitters. Dopamine-β-hydroxylase catalyses the synthesis of noradrenaline [1]. Nepicastat (SYN-117) HCl is a potent and selective dopamine-β-hydroxylase inhibitor. (R)-Nepicastat exhibited 2-3 fold less potent than nepicastat [1]. In beagle dogs and spontaneously hypertensive rats, nepicastat reduced noradrenaline in a dose-dependent way and increased dopamine and dopamine/noradrenaline ratio in cerebral cortex, left ventricle and the artery. In beagle dogs, nepicastat (2 mg/kg) significantly reduced noradrenaline by 52% and increased dopamine by 646% and dopamine/noradrenaline ratio in plasma [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0634
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.