Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%

Catalog No. AAH6305003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6305003 1 g
AAH6305006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6305003 Supplier Thermo Scientific Chemicals Supplier No. H6305003
Add to Cart

CAS: 148083-64-1 | C27H26N4O8 | 534.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148083-64-1
Molecular Formula C27H26N4O8
Molecular Weight (g/mol) 534.53
MDL Number MFCD00273454
InChI Key OENCMORJQAUAAJ-FQKVKQEKNA-N
Synonym fmoc-lys dnp-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-2,4-dinitrophenyl amino hexanoic acid, fmoc-l-lys dnp-oh, n-fmoc-n'-2,4-dinitrophenyl-l-lysine, fmoc-lys dnp, 2s-6-2,4-dinitrophenyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2,4-dinitrophenyl-l-lysine
PubChem CID 44593781
IUPAC Name 6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES OC(=O)[C@H](CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Formula Weight 534.52
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.