Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-Carbobenzoxy-D-lysine 98.0+%, TCI America™
SDP

Catalog No. C334525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C334525G 25 g
C33455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C334525G Supplier TCI America Supplier No. C334525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34404-32-5
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00063120
InChI Key CKGCFBNYQJDIGS-STGVRZAANA-N
Synonym h-d-lys z-oh, n6-phenylmethoxy carbonyl-r-lysine, r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 2r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, n-e-z-d-lysine, ambotzhaa6320, nepsilon-cbz-d-lysine, d-lys z-oh, nepsilon-carbobenzoxy-d-lysine, n6-benzyloxycarbonyl-d-lysine
PubChem CID 1551330
IUPAC Name (2R)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
SMILES N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 251°C
Color White
Quantity 25 g
Formula Weight 280.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-Carbobenzoxy-D-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.