Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages
GSA_VA

Catalog No. TGP401025PA
Change view
Click to view available options
Flavor:
Fruit Punch
Lemon-Lime
Orange
Dose:
50g
75g
100g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Dose
TGP401025PA Lemon-Lime 75g
TGP401009PA Lemon-Lime 100g
TGP401074PA Lemon-Lime 50g
TGP401272PA Orange 50g
TGP401503P Fruit Punch 100g
TGP401223PA Orange 75g
TGP401576P Fruit Punch 50g
TGP401526P Fruit Punch 75g
TGP401207PA Orange 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. TGP401025PA Supplier Thermo Scientific™ Supplier No. 401025P
This item is not returnable. View return policy

Screen for diabetes mellitus, gestational diabetes and hypoglycemia with these glucose tolerance test beverages.

Screen for diabetes mellitus, gestational diabetes and hypoglycemia with Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages. Beverages are noncarbonated, caffeine-free and available in 50, 75 and 100g concentrations.

  • Noncarbonated beverages provide easier consumption, a reduction of digestive discomfort and a more palatable taste
  • Graduated bottles assure proper dose every time
  • Kosher-certified

Ordering Alerts:

  • Plastic bottles packaged in cases of 24 bottles; four six-packs per case

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Kosher
Content And Storage 15°C to 30°C
Detectable Analytes None
Type Reagent
Dose 75g
Flavor Lemon-Lime
Packaging Type 24 Bottles/Cs.
Quantity 10 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.