Learn More
Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages

Description
Screen for diabetes mellitus, gestational diabetes and hypoglycemia with Thermo Scientific™ NERL™ Trutol™ Glucose Tolerance Test Beverages. Beverages are noncarbonated, caffeine-free and available in 50, 75 and 100g concentrations.
- Noncarbonated beverages provide easier consumption, a reduction of digestive discomfort and a more palatable taste
- Graduated bottles assure proper dose every time
- Kosher-certified
Ordering Alerts:
- Plastic bottles packaged in cases of 24 bottles; four six-packs per case
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Kosher |
| Content And Storage | 15°C to 30°C |
| Detectable Analytes | None |
| Type | Reagent |
| Dose | 75g |
| Flavor | Fruit Punch |
| Packaging Type | 24 Bottles/Cs. |
| Quantity | 10 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.