Learn More
TARGETMOL CHEMICALS INC NCGC00135472 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T336135MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. NCGC00135472 (DRV1 (GPR32) agonist C2A) is a human Resolvin DI receptor DRV1 agonist with pro-catabolic properties that activates the human soluble protein D1 receptor DRV1/GPR32 receptor in beta-blocker and cAMP assays with EC50s of 0.37 uM and 0.05 uM respectively.NCGC00135472 induces in cells overexpressing recombinant DRV1 rapid impedance changes that stimulate phagocytosis of serum-treated zymoglycans. purity: 99%
Catalog No. 50-258-0296
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.