Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nesprin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578438 Supplier Invitrogen™ Supplier No. PA578438
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: NIH-3T3 Predicted Reactivity: Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a nuclear outer membrane protein that binds cytoplasmic F-actin. This binding tethers the nucleus to the cytoskeleton and aids in the maintenance of the structural integrity of the nucleus. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nesprin 2
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene SYNE2
Gene Accession No. Q6ZWQ0, Q8WXH0
Gene Alias 6820443O06Rik; AW546258; D12Ertd777e; EDMD5; KASH domain-containing protein 2; KASH2; KIAA1011; MGC96690; mKIAA1011; Nesp2; Nesp2g; nesprin 2; nesprin-2; NUA; NUANCE; nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; polytrophin; Protein NUANCE; spectrin repeat containing nuclear envelope protein 2; spectrin repeat containing, nuclear envelope 2; synaptic nuclear envelope 2; synaptic nuclear envelope protein 2; synaptic nuclei expressed gene 2; SYNE2; Syne-2; TROPH
Gene Symbols SYNE2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse Nesprin 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23224, 319565
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.