Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nessler Tube Brushes
SCI_ED

Catalog No. S30435A
Change view
Click to view available options
Color:
Black
Length (English):
12 in.
20 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English)
S30435A Black 12 in.
S30435B Black 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30435A Supplier C&A Scientific Co Inc Supplier No. BR621A/EMD

For low-form Nessler tubes.

  • Radial, double-fan tip
  • Full bristles make complete contact with walls and bottom of cylinder
  • Galvanized wire handle

Specifications

Product Type Nessler Tube Brush
Bristle Type Radial, Double Fan Tip
Handle Material Galvanized Wire
Color Black
For Use With (Application) For low-form nessler tubes
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.