Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEU2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546722 Supplier Invitrogen™ Supplier No. PA546722
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ANVTRLCQVT STDHGRTWSS PRDLTDAAIG PAYREWSTFA VGPGHCLQLH Sequence homology: Dog: 77%; Human: 100%; Pig: 79%.

This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEU2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NEU2
Gene Accession No. Q9Y3R4
Gene Alias Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); SIAL2; sialidase 2 (cytosolic sialidase); sialidase-2
Gene Symbols NEU2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NEU2 (aa 119-168).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.