Learn More
Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (7.1), PE
Shop All Thermo Scientific Products

Description
This antibody recognizes an extracellular epitope of NG2, the melanoma-associated chondroitin sulfate proteoglycan 4 of Mw approximately 220-300 kDa. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
Specifications
Specifications
| Antigen | Neural/Glial Antigen 2 (NG2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 7.1 |
| Concentration | 0.055 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | Cspg4 |
| Gene Accession No. | Q6UVK1 |
| Gene Alias | 4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.