Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Neural Stem Cell Marker Antibody Pack
SDP

Catalog No. NBP142826 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142826 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142826 Supplier Novus Biologicals™ Supplier No. NBP142826

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Includes

  • 1.NB100-2322: HMGB1/HMG-1 Antibody
  • 2.NB110-37235: SOX2 Antibody
  • 3.NB300-266: Nestin Antibody (10C2)
  • 4.NBP1-05197: GFAP Antibody (5C10)

For Research applications

Neural Stem Cell Marker specifically detects Neural Stem Cell Marker in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Neural Stem Cell Marker
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Immunogen NB100-2322 - A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429]. NB110-37235 - A synthetic peptide made to the N-terminal region of human SOX2 protein (within residues 1-100). [Swiss-Prot# P48431]. MAB1195 - Neuron-specific beta-III Tubulin MAb (Clone TuJ-1). NBP1-05197 - A preparation of purified pig spinal cord GFAP. NB300-266 - A fragment of the human nestin protein corresponding to residues 1464-1614.
Purification Method Affinity Purified
Quantity 5 Vials
Research Discipline Neuronal Stem Cell Markers, Stem Cell Markers
Test Specificity NB100-2322: HMGB1 - Oligodendrocyte Marker NB110-37235: SOX2 - Embryonic Stem Cell Marker MAB1195: Neuron-specific beta-III Tubulin MAb (Clone TuJ-1) NBP1-05197: GFAP - Astrocyte Marker NB300-266: Nestin - Cytoskeleton-Intermediate Filaments Marker
Includes 1.NB100-2322: HMGB1/HMG-1 Antibody
2.NB110-37235: SOX2 Antibody
3.NB300-266: Nestin Antibody (10C2)
4.NBP1-05197: GFAP Antibody (5C10)
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.