Learn More
Worthington Biochemical Corporation NEURAMINIDASE
SDP
Supplier:  Worthington Biochemical Corporation LS004762
Encompass_Preferred
Chromatographically purified. A lyophilized powder containing 50% (w/w) sucrose. Contaminating proteolytic activity 0.1% using trypsin as the standard.Store at 2-8°C. PROTECT FROM MOISTURE.
Catalog No. NC2218581
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.