Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuraminidase, Clostridium perfringens, 50% w/w sucrose, Thermo Scientific Chemicals

Catalog No. AAJ60711CPL
Change view
Click to view available options
Quantity:
10 U
5 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60711CPL 5 U
AAJ60711LR 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60711CPL Supplier Thermo Scientific Chemicals Supplier No. J60711CPL
Add to Cart

A hydrolytic enzyme that removes sialic acid from mucoproteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-67-6
MDL Number MFCD00131711
Synonym Acyl-neuraminyl hydrolase,EC 3.2.1.18

Specifications

Packaging Plastic bottle
Quantity 5 U
Concentration or Composition (by Analyte or Components) 50% w/w sucrose
Physical Form Lyophilized Powder
Chemical Name or Material Neuraminidase
EINECSNumber 232-624-6
RTECSNumber QQ3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.