Learn More
ACROBiosystems Neuraminidase NA 1mg
Supplier:  ACROBiosystems NEEV52451MG
Encompass_Preferred
Influenza B [Austria/1359417/2021] Neuraminidase (NA) Protein His Tag (NEE-V5245) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Leu 466 (Accession # EPI_ISL_983345 GISAID).
Catalog No. 50-253-4378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.