Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NEUROBIOTIN(R) [Tracer]
SDP

Catalog No. NB397737 Shop All Novus Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397737 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397737 Supplier R&D Systems™ Supplier No. SP112020NB

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

  • An amino derivative of biotin designed for use as an intracellular label for cells, specifically neurons, to visualize neural architecture and identify gap junction coupling.
    • Versatile Preparation Use: Compatible with in vivo, whole mounts, slice preparations, and cultured cells.
    • Flexible Delivery Methods: Can be delivered via intracellular electrodes, microinjection, cut-loading, or scrape-loading.
    • Detection: Detectable using avidin or streptavidin systems via chromogenic or fluorescence visualization methods.
    • Key Advantage: Superior solubility compared to biocytin and other neuronal labels.

Specifications

Product Type Tracer
Quantity 20 mg
For Use With (Application) Bioimaging
Product Line Novus
Target Specificity NEUROBIOTIN(R)
Content And Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.