Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurocan Monoclonal Antibody (650.24)
GREENER_CHOICE

Catalog No. 01674944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-944 Supplier Invitrogen™ Supplier No. MA15843
  • $640.00
    $505.00 / Each of 1

    Save $135.00

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-5843 detects Neurocan in rat samples. MA1-5843 reacts with neurocan core protein, and also reacts with neurocan C fragment. MA1-5843 has been successfully used in Western blot, immunocytochemistry and immunofluorescence procedures. The MA1-5843 immunogen is embryonic rat brain proteoglycans.

Neurocan is a gene encoding a large chondroitin sulfate proteoglycan, predominantly expressed in the central nervous system. Located on chromosome 19 in humans, neurocan is part of the lectican family of proteoglycans, integral to the extracellular matrix where they modulate neuronal cell adhesion and migration. This proteoglycan is essential in the formation of the neural network during development and in maintaining the plasticity of the extracellular matrix in adult brains. Neurocan undergoes proteolytic processing, resulting in various fragments that influence neuronal growth and synapse stabilization. Altered neurocan expression has been linked to several neurological conditions such as schizophrenia, bipolar disorder, and Alzheimer's disease, likely due to its role in synaptic function and integrity of the neural matrix. Understanding neurocan's exact roles in neurological health and disease could illuminate pathways for potential therapeutic interventions targeting matrix remodeling and neuronal connectivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurocan
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 650.24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Ncan
Gene Accession No. O14594, P55066, P55067
Gene Alias 245 kDa early postnatal core glycoprotein; C230035B04; Chondroitin sulfate proteoglycan 3; chondroitin sulfate proteoglycan 3 (neurocan); chondroitin sulfate proteoglycan 3, related sequence; Cspg3; Cspg3-rs; NCAN; NEUR; neurocan; neurocan core protein; neurocan proteoglycan; TGFbeta-induced transcript, brain; Tgfbit
Gene Symbols Ncan
Host Species Mouse
Immunogen Embryonic rat brain proteoglycans from P3X cells.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13004, 1463, 58982
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.