Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD1 Mouse anti-Human, Clone: 3E10F1, Proteintech
SDP

Catalog No. p-7213359 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7108 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7108 Supplier Proteintech Group Inc Supplier No. 666911IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NeuroD1 encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.

Specifications

Antigen NEUROD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3E10F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NEUROD1
Gene Accession No. Q13562
Gene Alias BETA2, BHF 1, bHLHa3, NEUROD, NEUROD1, neurogenic differentiation 1
Gene Symbols NEUROD1
Host Species Mouse
Immunogen NEUROD1 Fusion Protein Ag27606
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4760
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.