Learn More
Invitrogen™ NEUROD4 Monoclonal Antibody (OTI1B7)

Description
NEUROD4 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | NEUROD4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NEUROD4 |
| Gene Accession No. | Q9HD90 |
| Gene Alias | AI846749; ath3; ATH-3; Atoh3; atonal homolog 3; atonal-like 3; atonal-like protein 3; bHLH domain; bHLHa4; Class A basic helix-loop-helix protein 4; helix-loop-helix protein mATH-3; mATH3; MATH-3; NeuroD4; neurod4 protein; neurogenic differentiation 4; neurogenic differentiation factor 4; neuronal basic helix-loop-helix transcription factor Ath3; neuronal differentiation 4; protein atonal homolog 3; zath3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.