Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeuroD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540540 Supplier Invitrogen™ Supplier No. PA540540
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MCRKFSRECE DQKQIKKPES FSKQIVLRGK SIKRAPGEET EKEEEEEDRE Sequence homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 79%; Rat: 100%.

NEUROD6 contains 1 basic helix-loop-helix (bHLH) domain. It activates E box-dependent transcription in collaboration with TCF3/E47 and may be a trans-acting factor involved in the development and maintenance of the mammalian nervous system. It transactivates the promoter of its own gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroD6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NEUROD6
Gene Accession No. Q96NK8
Gene Alias ATH2; Atoh2; atonal homolog 2; bHLHa2; brain my051 protein; class A basic helix-loop-helix protein 2; Helix-loop-helix protein mATH-2; MATH2; Math-2; My051; Neurod6; neurogenic differentiation 6; neurogenic differentiation factor 6; neuronal differentiation 6; Nex; Nex1; Nex1m; protein atonal homolog 2; Protein NEX-1; RGD1562793
Gene Symbols NEUROD6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NEUROD6 (aa 17-66).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63974
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.