Learn More
STEMCELL Technologies Neurodazine, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73292
Encompass
Neurodazine is a cell-permeable tri-substituted imidazole and neurogenic agent (Williams et al. 2007).DIFFERENTIATION· Induces neurogenesis of C2C12 myoblasts as well as mature human muscle cells, marked by upregulation of neural genes (Williams et al. 2007; Williams et al. 2008). · Induces neurogenesis and prevents astrocyte differentiation of P19 mouse embryonic carcinoma cells (Kim et al.).· Induces neuronal differentiation of human (SH-SY5Y) and mouse (Neuro-2a) neuroblastoma cells, and mouse NIH3T3 fibroblast cells (Halder et al.).
Catalog No. 50-197-6055
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.