Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurogenin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580851 Supplier Invitrogen™ Supplier No. PA580851
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NEUROG2.

Neurogenin-2 is a member of the neurogenin subfamily of basic helix-loop-helix (bHLH) transcription factor genes that play an important role in neurogenesis from migratory neural crest cells. In the course of mouse neurogenesis, Ngn2 and Ngn1 are expressed in distinct progenitor populations residing in the central and peripheral nervous systems. Mutation analyses demonstrate that Ngn1 is required for the determination of neuronal precursors for proximal cranial sensory ganglia and that Ngn2 is necessary for the determination of precursors for epibranchial placode-derived sensory neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin 2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NEUROG2
Gene Accession No. Q9H2A3
Gene Alias Ath4a; ATOH4; atonal homolog 4; bHLHa8; Class A basic helix-loop-helix protein 8; helix-loop-helix protein mATH-4A; mATH4A; NEUROG2; neurogenin 2; neurogenin2; neurogenin-2; NGN2; NGN-2; Protein atonal homolog 4
Gene Symbols NEUROG2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human NEUROG2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.