Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurogenin 2 Recombinant Superclonal™ Antibody (15HCLC)

Catalog No. PI711595 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711595 Supplier Invitrogen™ Supplier No. 711595
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Dog, Bovine and Rabbit Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Neurogenin-2 is a member of the neurogenin subfamily of basic helix-loop-helix (bHLH) transcription factor genes that play an important role in neurogenesis from migratory neural crest cells. In the course of mouse neurogenesis, Ngn2 and Ngn1 are expressed in distinct progenitor populations residing in the central and peripheral nervous systems. Mutation analyses demonstrate that Ngn1 is required for the determination of neuronal precursors for proximal cranial sensory ganglia and that Ngn2 is necessary for the determination of precursors for epibranchial placode-derived sensory neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin 2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 15HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene NEUROG2
Gene Accession No. P70447, Q9H2A3
Gene Alias Ath4a; ATOH4; atonal homolog 4; bHLHa8; Class A basic helix-loop-helix protein 8; helix-loop-helix protein mATH-4A; mATH4A; NEUROG2; neurogenin 2; neurogenin2; neurogenin-2; NGN2; NGN-2; Protein atonal homolog 4
Gene Symbols NEUROG2
Host Species Rabbit
Immunogen Protein corresponding to Human NEUROG2 (1-272).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11924, 63973
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.