Learn More
Enzo Life Sciences Neurokinin A (1mg). CAS: 86933-74-6
SDP
Supplier:  Enzo Life Sciences BMLP5050001
Encompass_Preferred
Endogenous ligand for NK2 receptor. Identical to substance K and neuromedin L. Alternative name: Neuromedin L, Substance K. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1356
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.