Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Neurokinin A Tachykinin Peptide, Solid, 1133.3 Da, 5 mg

Catalog No. 502707810
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7810 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7810 Supplier Abcam Supplier No. AB1201855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity tachykinin peptide for research applications.

  • Molecular weight: 1,133.3 Da
  • Purity: >95%
  • Form: Solid
  • Composition: 100% Neurokinin A
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Neurokinin A
Substrate Properties Peptide-based Substrate
Shipping Condition Ambient
Label or Dye Unconjugated
Content And Storage Store at -20°C.
Form Solid
Quantity 5 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.