Learn More
Medchemexpress LLC HY-P0242A 1mg Medchemexpress, Neurokinin B (TFA) CAS:101536-55-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0242A1MG
Encompass_Preferred
Medchemexpress, HY-P0242A 1mg Neurokinin B (TFA) CAS:101536-55-4 Neurokinin B TFA belongs to the tachykinin family of peptides. Neurokinin B binds a family of GPCRs-including neurokinin receptor 1 (NK1R), NK2R, and NK3R-to mediate their biological effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4073