Learn More
CPC Scientific H-Leu-Ser-Trp-Asp-Leu-Pro-Glu-Pro-Arg-Ser-Arg-Ala-Gly-Lys-Ile-Arg-Val-His-Pro-Arg-Gly-Asn-Leu-Trp-Ala-Thr-Gly-His-Phe-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP032B
Encompass
ONE-LETTER SEQUENCE: LSWDLPEPRSRAGKIRVHPRGNLWATGHFM-NH2MOLECULAR FORMULA: C157H243N51O38S1MOLECULAR WEIGHT:3485.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: NeuropeptidesREFERENCES: N. Minamino et al., BBRC, 130, 685 (1985)
Catalog No. 50-210-9916
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.