Learn More
Abcam Neuromedin B (Porcine), Bombesin-Like Peptide, 1 mg
Description
- Molecular weight: 1132.3 Da
- Purity: >90%
- Composition: 100% Neuromedin B (porcine)
- Form: Solid
- CAS Number: 87096-84-2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Neuromedin B |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Substrate Properties | Peptide-based Substrate |
| Quantity | 1 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.